Product Details

SNP ID
rs4174648
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:38585389 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCATCCCCACCCACAGCTGCCCCCT[A/C]TCTCCTGATCTCATTCGCAATGAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Tmem39a PubMed Links

Gene Details

Gene
Tmem39a
Gene Name
transmembrane protein 39a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205286.1 1180 Silent Mutation NP_001192215.1
NM_001205287.1 1180 Silent Mutation NP_001192216.1
NM_026407.3 1180 Silent Mutation NP_080683.2

View Full Product Details