Product Details
- SNP ID
-
rs13480757
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:108360797 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCAAAATAATTTTTCAGAAGTTA[A/G]TTGCATGTTTTATCATGGATAGACG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pawr
PubMed Links
Gene Details
- Gene
- Pawr
- Gene Name
- PRKC, apoptosis, WT1, regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_054056.2 |
|
Intron |
|
|
NP_473397.1 |
View Full Product Details