Product Details

SNP ID
rs4161630
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:11127166 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CATATGCATAGAGAAGTTTTAAAAT[A/G]TAAGTTGCCTCACATGCAGAAAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Txndc11 PubMed Links

Gene Details

Gene
Txndc11
Gene Name
thioredoxin domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029582.2 Intron NP_083858.1
NM_134105.2 Intron NP_598866.1
Gene
Zc3h7a
Gene Name
zinc finger CCCH type containing 7 A
There are no transcripts associated with this gene.

View Full Product Details