Product Details

SNP ID
rs13480841
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:4704211 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTGCGCCCTGATCGAAGGCTCGCTC[A/G]AAGAACAGGGCGCCCACTGCGATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Uqcr10 PubMed Links

Gene Details

Gene
Uqcr10
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197979.2 134 Silent Mutation NP_932096.1
Gene
Zmat5
Gene Name
zinc finger, matrin type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026015.2 134 Intron NP_080291.1

View Full Product Details