Product Details

SNP ID
rs13483220
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:14805677 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GATATTTAATGTATTAAGGTAATGT[A/G]ATTTCTAATTTCACTGAACTTTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Taf4b PubMed Links

Gene Details

Gene
Taf4b
Gene Name
TAF4B RNA polymerase II, TATA box binding protein (TBP)-associated factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100449.1 Intron NP_001093919.1

View Full Product Details