Product Details

SNP ID
rs13483341
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:46287401 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGCAGTCTTCCTCTTCTCTGC[C/T]TTCTCTGCCTTTTCCTTTATCTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ccdc112 PubMed Links

Gene Details

Gene
Ccdc112
Gene Name
coiled-coil domain containing 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160399.1 1399 Silent Mutation NP_001153871.1
Gene
Pggt1b
Gene Name
protein geranylgeranyltransferase type I, beta subunit
There are no transcripts associated with this gene.

View Full Product Details