Product Details
- SNP ID
-
rs13482014
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:111487036 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGCGCTCCCGCTCGCTCTCTCTCT[A/C]TATATATATATATATGCTATCATAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Gpbp1
PubMed Links
Gene Details
- Gene
- Gpbp1
- Gene Name
- GC-rich promoter binding protein 1
View Full Product Details