Product Details
- SNP ID
-
rs3692409
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:160811177 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGGGGAACAACAGTCAGGACAAG[A/G]ACTGGTTTTAAGATCAAAGGGACAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Zhx3
PubMed Links
Gene Details
- Gene
- Zhx3
- Gene Name
- zinc fingers and homeoboxes 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177263.3 |
|
Intron |
|
|
NP_796237.2 |
View Full Product Details