Product Details

SNP ID
rs13476293
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:188263219 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACGCCACGTGTGGATTCCCAGGCCC[A/T]AGCACTTTTTGTCGTAGCCCGGTGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Ush2a PubMed Links

Gene Details

Gene
Ush2a
Gene Name
Usher syndrome 2A (autosomal recessive, mild)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021408.3 382 Silent Mutation NP_067383.3

View Full Product Details