Product Details

SNP ID
rs32147763
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:43458888 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTTCTTGTGGAACAGTGGGT[G/T]TTGCAGATCAACCCTGTCCAGCTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Nck2 PubMed Links

Gene Details

Gene
Nck2
Gene Name
non-catalytic region of tyrosine kinase adaptor protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010879.3 Intron NP_035009.3

View Full Product Details