Product Details
- SNP ID
-
rs13483228
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:16786338 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAGACTGACTCTGTCCATATGGG[A/G]AAAAAAACACCACAGATTGCTAGTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cdh2
PubMed Links
Gene Details
- Gene
- Cdh2
- Gene Name
- cadherin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007664.4 |
|
Intron |
|
|
NP_031690.3 |
View Full Product Details