Product Details

SNP ID
rs13478130
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:20760935 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATAAATATAAGAATTGCACAATACA[G/T]TCTGATGTAAACACTTCTTTCCCAG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
Phtf2 PubMed Links

Gene Details

Gene
Phtf2
Gene Name
putative homeodomain transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172992.3 2568 UTR 3 NP_766580.3

View Full Product Details