Product Details
- SNP ID
-
rs13480998
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:43869506 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTTGTTTTCTCAGCCTAGTCCA[C/T]GAAGATGGAAGATGGCAAAGGTCTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Adra1b
PubMed Links
Gene Details
- Gene
- Adra1b
- Gene Name
- adrenergic receptor, alpha 1b
View Full Product Details