Product Details
- SNP ID
-
rs6291204
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:165394046 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCACATGTGATACATAGACGCA[A/G]TGAGATACAGTTCATCTGTAAAGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dcaf6
PubMed Links
Gene Details
- Gene
- Dcaf6
- Gene Name
- DDB1 and CUL4 associated factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028759.1 |
|
Intron |
|
|
NP_083035.1 |
View Full Product Details