Product Details
- SNP ID
-
rs13477745
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:65386538 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGCTTTGATCCTCAGTTTCCTC[A/T]TTTGTCAAACAGGATTCAGGGCATT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Astn2
PubMed Links
Gene Details
- Gene
- Astn2
- Gene Name
- astrotactin 2
View Full Product Details