Product Details
- SNP ID
-
rs13481255
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:117732288 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGCAGCGTGGGGGAAGCCTCCCA[A/G]CAGTGGCAGTGGGTGGGATCATCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tnrc6c
PubMed Links
Gene Details
- Gene
- Tnrc6c
- Gene Name
- trinucleotide repeat containing 6C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198022.2 |
3569 |
Missense Mutation |
|
|
NP_932139.2 |
View Full Product Details