Product Details
- SNP ID
-
rs13480683
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:85647373 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTAAGCTCCTCCTCCTCTGGAG[C/T]AAAGCTTGAACAATAGAAAGGCCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Btbd11
PubMed Links
Gene Details
- Gene
- Btbd11
- Gene Name
- BTB (POZ) domain containing 11
View Full Product Details