Product Details
- SNP ID
-
rs13478684
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29410953 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTAATGGCACCATTTTCATTTGC[A/G]CTTAGTTCACTGTCAGGGTATATAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ccdc136
PubMed Links
Gene Details
- Gene
- Ccdc136
- Gene Name
- coiled-coil domain containing 136
View Full Product Details