Product Details
- SNP ID
-
rs13483141
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:85336962 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTCAGGCACCAGGAAAAAAGCTC[C/T]GAGTAGCAGTTTTCTTCCAACTCCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Camkmt
PubMed Links
Gene Details
- Gene
- Camkmt
- Gene Name
- calmodulin-lysine N-methyltransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_028576.2 |
|
Intron |
|
|
NP_082852.1 |
View Full Product Details