Product Details

SNP ID
rs13481613
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:100912888 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGGACGCCGGTGTGCTCTTTGAACT[A/G]CTCCCTTGACTGAAGTCATCGGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ccdc88c PubMed Links

Gene Details

Gene
Ccdc88c
Gene Name
coiled-coil domain containing 88C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026681.4 6104 Silent Mutation NP_080957.2
Gene
Gpr68
Gene Name
G protein-coupled receptor 68
There are no transcripts associated with this gene.

View Full Product Details