Product Details
- SNP ID
-
rs13476910
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:169752280 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCCTTTTAAAATCATCTTTAAT[C/T]GAATCGTATTGTGATTACTTCCGCG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tshz2
PubMed Links
Gene Details
- Gene
- Tshz2
- Gene Name
- teashirt zinc finger family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080455.2 |
|
Intron |
|
|
NP_536703.2 |
View Full Product Details