Product Details

SNP ID
rs13477683
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:47049208 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCCCTTCTCCACCAGCTGCTG[C/T]GCCACATTCAGCTTCCCAAGGAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Anks6 PubMed Links

Gene Details

Gene
Anks6
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024136.1 722 Silent Mutation NP_001019307.1

View Full Product Details