Product Details

SNP ID
rs4222275
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:37410413 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTTCCAGACGGCCCGTCCTGC[T/G]TCTATCCCCATTTAAAGGAACGTGC
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
Coa5 PubMed Links

Gene Details

Gene
Coa5
Gene Name
cytochrome C oxidase assembly factor 5
There are no transcripts associated with this gene.

Gene
Inpp4a
Gene Name
inositol polyphosphate-4-phosphatase, type I
There are no transcripts associated with this gene.

View Full Product Details