Product Details
- SNP ID
-
rs3673679
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59876487 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGCCCAGAAAGGAGTCAGCAGG[T/C]GTGAATATTTGTTGCCAATATGAAA
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Nt5m
PubMed Links
Gene Details
- Gene
- Nt5m
- Gene Name
- 5',3'-nucleotidase, mitochondrial
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_134029.2 |
1260 |
UTR 3 |
|
|
NP_598790.1 |
View Full Product Details