Product Details
- SNP ID
-
rs4230094
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:113095757 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCATCTTTTTCACTTACATTGCA[G/C]GAGTCCTTTCCACCACGGAGGTCCC
- Phenotype
-
- Polymorphism
- G/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Cdc20b
PubMed Links
Gene Details
- Gene
- Cdc20b
- Gene Name
- cell division cycle 20B
- Gene
- Gzma
- Gene Name
- granzyme A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010370.2 |
648 |
Silent Mutation |
|
|
NP_034500.1 |
View Full Product Details