Product Details

SNP ID
rs4151956
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.X:136214619 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGAGAAGGAAAAGATTTCTTCGTGG[A/G]TTGATCACTAACGAGGACAAGAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Bex1 PubMed Links

Gene Details

Gene
Bex1
Gene Name
brain expressed gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009052.2 Intron NP_033078.2
Gene
Tceal7
Gene Name
transcription elongation factor A (SII)-like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127169.1 Intron NP_001120641.1

View Full Product Details