Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUC15
Entrez Gene ID
102134216
Gene Name
mucin 15, cell surface associated
Gene Aliases
-
Chromosome Location
Chr.14: 40925197 - 40939084 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005578365.2 XP_005578422.1 3 - 4 1558 63

View Full Product Details