Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD13
Entrez Gene ID
102122850
Gene Name
abhydrolase domain containing 13
Gene Aliases
-
Chromosome Location
Chr.17: 90716203 - 90732138 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586223.2 XP_005586280.1 1 - 2 273 84

View Full Product Details