Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL14
Entrez Gene ID
102121072
Gene Name
ADP ribosylation factor like GTPase 14
Gene Aliases
-
Chromosome Location
Chr.2: 63080312 - 63081636 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005546258.2 XP_005546315.1 1 - 2 150 87

View Full Product Details