Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H4orf33
Entrez Gene ID
102121777
Gene Name
chromosome 5 open reading frame, human C4orf33
Gene Aliases
-
Chromosome Location
Chr.5: 128078988 - 128100362 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555919.2 XP_005555976.1 6 - 7 956 79
XM_015450837.1 XP_015306323.1 7 - 8 1091 79

View Full Product Details