Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BST2
Entrez Gene ID
102128665
Gene Name
bone marrow stromal cell antigen 2
Gene Aliases
-
Chromosome Location
Chr.19: 17733208 - 17735857 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005588381.2 XP_005588438.1 1 - 2 372 71

View Full Product Details