Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
102144827
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
-
Chromosome Location
Chr.7: 69923346 - 69932613 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560485.2 XP_005560542.1 2 - 3 335 102

View Full Product Details