Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG2
Entrez Gene ID
102145250
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 2
Gene Aliases
-
Chromosome Location
Chr.10: 14568110 - 14711597 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567399.2 XP_005567456.1 2 - 3 466 97

View Full Product Details