Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNIP2
Entrez Gene ID
102140778
Gene Name
TNFAIP3 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 2794781 - 2812713 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554316.1 XP_005554373.1 1 - 2 339 134
XM_005554317.1 XP_005554374.1 1 - 2 339 134

View Full Product Details