Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRD1
Entrez Gene ID
101865257
Gene Name
leucine rich repeats and death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 119805621 - 119832013 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283368.1 NP_001270297.1 4 - 5 2481 99
XM_015447256.1 XP_015302742.1 5 - 6 1218 99
GenBank mRNA AB168315.1 - 3 - 4 1599 99
AB169234.1 - 4 - 5 2481 99

View Full Product Details