Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC13
Entrez Gene ID
102134679
Gene Name
WAP four-disulfide core domain 13
Gene Aliases
-
Chromosome Location
Chr.10: 77387433 - 77392189 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005569165.2 XP_005569222.1 1 - 2 437 110

View Full Product Details