Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102135713
Entrez Gene ID
102135713
Gene Name
serine protease inhibitor Kazal-type 6
Gene Aliases
SPINK6
Chromosome Location
Chr.6: 147449672 - 147461305 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015452105.1 XP_015307591.1 1 - 2 538 79

View Full Product Details