Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A5
Entrez Gene ID
102143071
Gene Name
solute carrier family 16 member 5
Gene Aliases
-
Chromosome Location
Chr.16: 72353897 - 72372687 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438823.1 XP_015294309.1 2 - 3 1472 85
XM_015438824.1 XP_015294310.1 3 - 4 749 85
XM_015438825.1 XP_015294311.1 3 - 4 712 85
XM_015438826.1 XP_015294312.1 3 - 4 518 85

View Full Product Details