Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4EBP1
Entrez Gene ID
102132583
Gene Name
eukaryotic translation initiation factor 4E binding protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 38838889 - 38871454 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563062.2 XP_005563119.1 2 - 3 575 76

View Full Product Details