Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD60
Entrez Gene ID
102126101
Gene Name
ankyrin repeat domain 60
Gene Aliases
-
Chromosome Location
Chr.10: 90109838 - 90120357 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005569684.2 XP_005569741.1 1 - 2 260 59

View Full Product Details