Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLECL1
Entrez Gene ID
102141146
Gene Name
C-type lectin like 1
Gene Aliases
-
Chromosome Location
Chr.11: 10083918 - 10104516 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570088.2 XP_005570145.1 2 - 3 1680 97

View Full Product Details