Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLTPD2
Entrez Gene ID
102128996
Gene Name
glycolipid transfer protein domain containing 2
Gene Aliases
-
Chromosome Location
Chr.16: 4843079 - 4845279 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582583.2 XP_005582640.1 1 - 2 110 90

View Full Product Details