Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCA1L
Entrez Gene ID
102121343
Gene Name
IQ motif containing with AAA domain 1 like
Gene Aliases
IQCA1P1
Chromosome Location
Chr.3: 183880302 - 183895143 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551202.2 XP_005551259.2 18 - 19 2444 101

View Full Product Details