Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102139127
Entrez Gene ID
102139127
Gene Name
alkaline phosphatase, placental-like
Gene Aliases
ALPPL2
Chromosome Location
Chr.12: 122215150 - 122218901 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574616.2 XP_005574673.1 7 - 8 1052 62

View Full Product Details