Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLEC10A
Entrez Gene ID
102131825
Gene Name
C-type lectin domain containing 10A
Gene Aliases
-
Chromosome Location
Chr.16: 7105989 - 7112738 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015437550.1 XP_015293036.1 7 - 8 1795 94

View Full Product Details