Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Casp7
- Entrez Gene ID
-
12369
- Gene Name
- caspase 7
- Gene Aliases
-
AI314680, CMH-1, ICE-IAP3, Mch3, caspase-7, mCASP-7
- Chromosome Location
-
Chr.19: 56396091 - 56442347 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
U67321.1 |
- |
|
829 |
|
Chr. - - [-] on Build |
100 |
View Full Product Details