The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008106.2 | NP_032132.1 | 4 - 5 | 771 | 87 |
GenBank mRNA | AF011389.1 | - | 4 - 5 | 757 | 87 |
AF190672.1 | - | 4 - 5 | 773 | 87 | |
AK149287.1 | - | 4 - 5 | 772 | 87 | |
BC014826.1 | - | 4 - 5 | 751 | 87 |