Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsf2
Entrez Gene ID
15500
Gene Name
heat shock factor 2
Gene Aliases
AI661205
Chromosome Location
Chr.10: 57486187 - 57513143 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008297.3 NP_032323.3 1 - 2 214 64
GenBank mRNA AK052270.1 - 1 - 2 113 64
AK053976.1 - 1 - 2 215 64
BC018414.1 - 1 - 2 136 64
X61754.1 - 1 - 2 248 64

View Full Product Details