Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsf2
Entrez Gene ID
15500
Gene Name
heat shock factor 2
Gene Aliases
AI661205
Chromosome Location
Chr.10: 57486187 - 57513143 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008297.3 NP_032323.3 6 - 7 718 154
GenBank mRNA AK052270.1 - 6 - 7 617 154
AK053976.1 - 6 - 7 719 154
BC018414.1 - 6 - 7 640 154
X61754.1 - 6 - 7 752 154

View Full Product Details