The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008373.1 | NP_032399.1 | 4 - 5 | 326 | 81 |
GenBank mRNA | X14045.1 | - | 4 - 5 | 326 | 81 |